Order Entry
Germany
ContactUsLinkComponent
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
Catalog # 20838.294
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  
undefined
2-Propanol, dehydrated (max. 0.01% H₂O) ≥99.8%, AnalaR NORMAPUR® analytical reagent
Catalog # 20838.294
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Colour value:
    Max. 10 APHA
  • Assay:
    Min. 99.8 %
  • Aldehydes:
    Max. 0.0002 %
  • Formaldehyde:
    Max. 0.0002 %
  • Ketones (as C3H6O):
    Max. 0.0050 %
  • Methanol:
    Max. 0.1 %
  • Residue on evaporation:
    Max. 0.0010 %
  • Water (K.F.):
    Max. 0.0100 %
  • Formaldehyde-sulphuric colouration:
    Max. 60.0 APHA
  • Acidity:
    Max. 0.0020 %
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.1 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.01 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Ni (Nickel):
    Max. 0.01 ppm
  • Pb (Lead):
    Max. 0.01 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.01 ppm

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674