2-Propanol
5 L
LBCL4561205
Marketsource
: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
- :67-63-0
- :(CH₃)₂CHOH
- :MFCD00011674
- :60.1 g/mol
- :13,05228
- :82 °C (1013 hPa)
- :–89 °C
- :0.786 g/cm³ (20 °C)
- :12 °C
- :Ambient
- :200-661-7
- :1219
- :3,II
- :01-2119457558-25
- Pk:5 L