Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-Octanol
Supelco® Organic Reference Standard, 1-Octanol
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, 1-Octanol
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    111-87-5
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H319 - H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002988
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Alcohol C₈; Capryl alcohol; Octyl alcohol
  • Melting point:
    –15 °C (lit.)
  • Formula:
    CH₃(CH₂)₇OH
  • Molecular weight:
    130,23
  • Density:
    0,827 g/ml at 25 °C (lit.)
  • Flash point:
    86,5 °C
  • Boiling point:
    196 °C (lit.)

Specifications

Related Information

Frequently Bought Together