1-Octadecanol, Supelco®
Catalog # 74723-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:270.5 g/mol
Merck Index:12,08960
Boiling Pt:336 °C (1013 hPa)
Melting Pt:56…57 °C
Density:0.805…0.815 g/cm³ (60 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:204-017-6
CAS Number:112-92-5
Formula:H₃C(CH₂)₁₆CH₂OH
MDL Number:MFCD00002823