Order Entry
Germany
ContactUsLinkComponent
1-Octadecanol 95%
1-Octadecanol 95%
Certificates
undefined
1-Octadecanol 95%
Certificates

Specification Test Results

  • Appearance:
    White Powder or flakes or pellets
  • Infrared spectrum:
    Conforms
  • Melting point:
    55 to 60 °C
  • GC:
    ≥94 %

Specification Test Results

Related Information