Order Entry
Germany
ContactUsLinkComponent
1-Hexadecanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
1-Hexadecanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Catalog # 1.00989.9024
Supplier:  Merck
CAS Number:  
1-Hexadecanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, SAFC®
Catalog # 1.00989.9024
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    20 kg
  • Seq:
    4241

Specifications

About this item

 :
 :242.45 g/mol
Merck Index:13,02037
Boiling Pt:305…330 °C (1013 hPa)
Melting Pt:46…52 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:253-149-0
CAS Number:36653-82-4
Formula:H₃C(CH₂)₁₄CH₂OH
MDL Number:MFCD00004760