1-Butanol
5 L
Catalog # LBCL4561206
Marketsource
Supplier: LABC LABORTECHNIK
Jump to:
Product Details & Documents
Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol
- CAS Number:71-36-3
- Formula:H₃C(CH₂)₂CH₂OH
- MDL Number:MFCD00002964
- Molecular Weight:74.12 g/mol
- Merck Index:13,01539
- Boiling Pt:117.6 °C (1013 hPa)
- Melting Pt:–89 °C
- Density:0.812 g/cm³ (20 °C)
- Flash Pt:30 °C
- Storage Temperature:Ambient
- EINECS:200-751-6
- UN:1120
- ADR:3,III
- REACH:01-2119484630-38
Specifications
Specifications
- Pk:5 L



