Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Diethyl ether, Supelco®

Flammable
Irritant
Supplier:   Merck
Diethyl ether, Supelco®
Supplier:  Merck
Certificates
Diethyl ether, Supelco®
Diethyl ether, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 51,30 €
    51,30 €
    1 * 1 mL
       51,30 €

  • Glass bottle
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 351,00 €
    351,00 €
    1 * 10 mL
       351,00 €