

Diethyl ether, anhydrous ≥99.0% contains inhibitor ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:34…35 °C (1.013 hPa)
Melting Pt:–116 °C (1.013 hPa)
Density:0.71 g/cm³ (20 °C)
Flash Pt:–45 °C (1.013 hPa)
Storage Temperature:Ambient
UN:1155
ADR:3,I
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:200-467-2
REACH:01-2119535785-29
:GHS07
:GHS02
Specification Test Results
- IR spectrum:complying
- Assay (GC):Min. 99.0 %
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.03 %
- Titrable Acid:Max. 0.0002 meq/g
- Carbonyl compounds (as CO):Max. 0.001 %
- Ethanol (GC):Max. 0.02 %
- Methanol (GC):Max. 0.02 %
- Peroxides (as H₂O₂):Max. 1 ppm
- APHA:Max. 10
- Assay BHT (GC):Min. 1 ppm
- Refractive Index:n20/D 1.3530 (lit.)
- Explosion Limit:36.5 %
- Contains:BHT as inhibitor