Diethyl ether stabilised Ph. Eur.
Supplier: VWR Chemicals
About this item
Appearance: Clear, colourless, volatile, very mobile liquid.
Caution: Not for use as Active Pharmaceutical Ingredient (API)
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
3 Options Available Below
Product Details & Documents
Appearance: Clear, colourless, volatile, very mobile liquid.
Caution: Not for use as Active Pharmaceutical Ingredient (API)
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Merck Index:12,03852
- Boiling Pt:35 °C (1013 hPa)
- Melting Pt:–123 °C
- Density:0.7135 g/cm³ (20 °C)
- Flash Pt:–40 °C
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
- Storage Temperature:Ambient
- EINECS:200-467-2
- REACH:01-2119535785-29
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 5L Aluminium bottle for liquids DIN 45 UN X (ZPACK8000430)
Packaging Information
23819_DIETHYL ETHER PH.EUR._BSE_REV1
BSE/TSE Certificates
Diethyl Ether Stabilised Ph. Eur.
Additional Product Details
P20_VWRC23819.298_SPEC_EN.pdf P20_VWRC23819.367_SPEC_EN.pdf P20_VWRC23819.460_SPEC_EN.pdf spec_VWRC23819.298 spec_VWRC23819.298
Product specification
More
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Specifications for Product Family
Specification Test Results
- Assay (Ionol/BHT):Passes test
- Appearance:Conforms (see CoA/CoS)
- Acidity:Passes test
- Relative density:0.714 - 0.716
- Distillation range:34.0 - 35.0 °C
- Acetone and aldehydes:Passes test
- Peroxides:Passes test
- Non-volatile matter:≤ 20 mg/l
- Unspecified impurities:≤ 0.05 %
- Total impurities:≤ 0.1 %
- Reporting threshold:≤ 0.03 %
- Water:≤ 2 g/l
- Residual solvents:Passes test
- As (Arsenic):≤ 0.1 ppm
- Cd (Cadmium):≤ 0.1 ppm
- Co (Cobalt):≤ 0.1 ppm
- Hg (Mercury):≤ 0.1 ppm
- Ni (Nickel):≤ 0.1 ppm
- Pb (Lead):≤ 0.1 ppm
- V (Vanadium):≤ 0.1 ppm
- Conforms to Ph.Eur.:Passes test
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



