



2,2,2-Trifluoroethanol 99.5% for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-89-8
:
Formula:C₂H₃F₃O
MDL Number:MFCD00004672
:100.04 g/mol
Boiling Pt:74…75 °C
Melting Pt:–44 °C
Density:1.391
Flash Pt:29 °C (84 °F)
EINECS:200-913-6
UN:1992
ADR:3,III
:GHS06
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient