Trifluoroacetic acid
Catalog # A0697.0100
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:114.02 g/mol
Merck Index:13,09754
Boiling Pt:71.8 °C (1013 hPa)
Melting Pt:–15 °C
Density:1.490 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-929-3
UN:2699
ADR:8,I
:GHS05
:GHS07
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169