
Perfluorohexane
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:355-42-0
:
Formula:C₆F₁₄
MDL Number:MFCD00000437
:338.04 g/mol
Boiling Pt:56 °C (1013 hPa)
Melting Pt:–4 °C
Density:1,68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-585-0
:GHS07