Trichloroacetic acid (TCA), EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
EINECS:200-927-2
UN:1839
ADR:8,II
:M-Clarity™ quality level = MQ300
Flash Pt:113 °C (closed cup)
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:1600 g/l (20 °C)
- Melting Point:54 - 56 °C
- Molar Mass:163.38 g/mol
- Bulk Density:900 kg/m3
- Boiling Point:197 °C
- Vapor Pressure:1 hPa (20 °C)
- Flash Point:110 °C
- Density:1.63 g/cm3 (20 °C)
- pH:- 1 (50 g/l, H2O, 20 °C)
- Ignition Point:711 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00807.0250_TDS_EN ProductPage_100807
Product specification
msds_100807