Trichloroacetic acid (TCA) ≥99.0%, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:197 °C (1013 hPa) (lit.)
Melting Pt:∼56 °C (lit.)
Density:1.61 g/cm³ (lit.)
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-927-2
UN:1839
ADR:8,II
:GHS05
:GHS07
:GHS09
Specification Test Results
- Purity:≥99.0%
- Residue after Ignition:≤0.03%
- Chloride:≤0.002%
- Nitrate:≤0.002%
- Phosphate:≤5ppm
- Sulfate:≤0.02%
- Heavy Metals:≤0.002%
- Iron:≤0.001%
- Substance Darkened by Sulfuric Acid:Passes