Trichloroacetic acid (TCA), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (1013 hPa)
Melting Pt:54…56 °C
Density:1.63 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
EINECS:200-927-2
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay:min. 99.0 %
- Clarity of Solution:passes test
- Chloride (Cl):max. 0.001 %
- Iron (Fe):max. 0.001 %
- Heavy Metals (as Pb):max. 0.002 %
- Insoluble Matter:max. 0.005 %
- Nitrate (NO3):max. 0.002 %
- Residue after Ignition:max. 0.02 %
- Substances Darkened by H2SO4:passes test
- Sulfate (SO4):max. 0.02 %
- Phosphate (PO4):max. 5