Trichloroacetic acid (TCA) ≥99% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:14,9627
Boiling Pt:196 °C
Melting Pt:54…58 °C
Density:1.629
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-927-2
UN:1839
ADR:8,II
:GHS05
:GHS08
:GHS09