Trichloroacetic acid (TCA) 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-03-9
:
Formula:Cl₃CCOOH
MDL Number:MFCD00004177
:163.39 g/mol
Merck Index:13,09700
Boiling Pt:196 °C (760 mmHg)
Melting Pt:52…58 °C
Density:1.62 g/cm³
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-927-2
UN:1839
ADR:8,II
:GHS07
:GHS05
:GHS09
Specification Test Results
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Residue after ignition:≤0.03 %
- Titration with NaOH:≥99.0 %
- Clarity of solution:Passes test
- Substance darkened by H2SO4:Passes test
- Chloride (Cl):≤20 ppm
- Nitrate (NO3):≤20 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤200 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤20 ppm
- Insoluble matter:≤0.01 %