ContactUsLinkComponent
Chloroform stabilised, Supelco®
  02487-5ML
 :  Merck
 :  
undefined
Chloroform stabilised, Supelco®
  02487-5ML
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
 :
 :119.38 g/mol
 :13,02160
 :61.7 °C (1013 hPa)
 :–63 °C
 :1.4832 g/cm³ (20 °C)
 :Ambient
 :200-663-8
 :1888
 :6.1,III
 :M-Clarity™ quality level = MQ100
 :05-2114614906-45
 :GHS06
 :GHS08
 :67-66-3
 :CHCl₃
 :MFCD00000826