Order Entry
Germany
ContactUsLinkComponent
Chloroform ≥99% stabilised, GPR RECTAPUR®
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99 %
  • Boiling point:
    59.5 - 61.5 °C
  • Density (20/4):
    1.478 - 1.489
  • Evaporation residue:
    Max. 10 ppm
  • Free chlorine:
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Water:
    Max. 100 ppm

Specification Test Results