Order Entry
Germany
ContactUsLinkComponent
Chloroform stabilised, SupraSolv® for GC-ECD/FID, for residue analysis, Supelco®
Toxic
Health
Chloroform stabilised, SupraSolv® for GC-ECD/FID, for residue analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Toxic
Health
Chloroform stabilised, SupraSolv® for GC-ECD/FID, for residue analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    8 g/L (20 °C)
  • Melting Point:
    -63 °C
  • Molar Mass:
    119.38 g/mol
  • Boiling Point:
    61 °C (1013 hPa)
  • Vapor Pressure:
    211 hPa (20 °C)
  • Refractive Index:
    1.45
  • Density:
    1.48 g/cm³ (20 °C)
  • pH:
    (H₂O) no data available
  • Ignition Point:
    982 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Frequently Bought Together