Tetrachloromethane, Supelco®
Glass bottle, 1 mL
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Carbon tetrachloride, TCT
- CAS Number:56-23-5
- Formula:CCl₄
- Molecular Weight:153.82 g/mol
- Merck Index:12,01864
- Boiling Pt:76.7 °C (1013 hPa)
- Melting Pt:–23 °C
- Density:1.5942 g/cm³ (20 °C)
- ADR:6.1,II
- :M-Clarity™ quality level = MQ200
- :Density: 1.5942 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-262-8
- UN:1846
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:1 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



