Order Entry
Germany
ContactUsLinkComponent
2-Chlorobutane for synthesis, Sigma-Aldrich®
2-Chlorobutane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-Chlorobutane for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1.0 g/l (25 °C)
  • Melting Point:
    -132 °C
  • Molar Mass:
    92.56 g/mol
  • Boiling Point:
    65 °C (1013 hPa)
  • Vapor Pressure:
    160 hPa (20 °C)
  • Flash Point:
    -10 °C
  • Refractive Index:
    1.395 (20 °C, 589 nm)
  • Explosion Limit:
    2.0 - 8.8 %(V)
  • Density:
    0.87 g/cm3 (20 °C)
  • pH:
    7 (H2O, 20 °C) Aqueous solution
  • Ignition Point:
    460 °C

Specification Test Results

Related Information