2-Chlorobutane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-86-4
:
Formula:C₄H₉Cl
MDL Number:MFCD00000871
:92.57 g/mol
Merck Index:14,1561
Boiling Pt:67…69 °C
Melting Pt:–141…–140 °C
Density:0.873
Flash Pt:–15 °C (5 °F)
Storage Temperature:Ambient
EINECS:201-151-7
UN:1127
ADR:3,II
:GHS02