Order Entry
Germany
ContactUsLinkComponent
 
1-Chlorododecane for synthesis, Sigma-Aldrich®
 :  Merck
 
1-Chlorododecane for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
     - 0.007 g/l (25 °C)
  • Melting Point:
    -9 °C
  • Molar Mass:
    204.78 g/mol
  • Boiling Point:
    124 °C (13 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    130 °C
  • Density:
    0.87 g/cm3 (20 °C)
  • Ignition Point:
    190 °C