


1,2-Dichloroethane 99+% for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-06-2
:
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963
:98.96 g/mol
Merck Index:14,3797
Boiling Pt:83 °C
Melting Pt:–35 °C
Density:1.256
Flash Pt:15 °C (59 °F)
Storage Temperature:Ambient
EINECS:203-458-1
UN:1184
ADR:3,II
:GHS02
:GHS08
:GHS07