Order Entry
Germany
ContactUsLinkComponent
Citric acid for synthesis, Sigma-Aldrich®
Irritant
Citric acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Irritant
Citric acid for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    1330 g/l (20 °C)
  • Melting Point:
    153 °C (decomposition)
  • Molar Mass:
    192.12 g/mol
  • Bulk Density:
    560 kg/m3
  • Boiling Point:
    200 °C (1013 hPa) (decomposition)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Density:
    1.665 g/cm3 (18 °C)
  • pH:
    1.7 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    345 °C

Specification Test Results

Related Information