Order Entry
Germany
ContactUsLinkComponent
 
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 
undefined
Ammonium acetate ≥98.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 

 

  • Assay:
    Min. 98.0 %
  • Appearance:
    Colourless crystals
  • pH (25°C; 5 %):
    6.7 - 7.3
  • Heavy metals (as Pb):
    Max. 2 ppm
  • Ignition residue (SO4):
    Max. 100 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Substances reducing KMnO4 (as HCOOH):
    Max. 50 ppm
  • Water:
    Max. 2.0 %
  • Cl (Chloride):
    Max. 5 ppm
  • NO3 (Nitrate):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 10 ppm
  • Ca (Calcium):
    Max. 10 ppm
  • Cd (Cadmium):
    Max. 0.5 ppm
  • Cu (Copper):
    Max. 0.5 ppm
  • Fe (Iron):
    Max. 2 ppm
  • Ni (Nickel):
    Max. 0.5 ppm
  • Pb (Lead):
    Max. 0.5 ppm
  • Zn (Zinc):
    Max. 0.5 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test