Ammonium acetate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Merck Index:14,495
Boiling Pt: Dec.
Melting Pt:∼114 °C (dec.)
Density:1.17
Flash Pt:136 °C (277 °F)
EINECS:211-162-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Form:Crystals or powder or crystalline powder
- Identification (FTIR):Conforms
- Ammonia as ammonium acetate:≥96.0% to ≤104.0%
- Water Content (Karl Fischer Titration):≤5.0%
- Appearance (Color):White