Ammonium acetate 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:631-61-8
:
Formula:H₃CCOONH₄
MDL Number:MFCD00013066
:77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1,17 g/cm³ (20 °C)
Flash Pt:136 °C
Storage Temperature:Ambient
EINECS:211-162-9
Specification Test Results
- Appearance:White Adhering crystals
- Infrared spectrum:Conforms
- Water:≤2 %
- Titration with HClO4:≥97.5 % (On anhydrous substance)