Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium acetate
Potassium acetate, Millipore®
Potassium acetate, Millipore®
:
Merck Millipore (Calbiochem)
Potassium acetate, Millipore®
:
Merck Millipore (Calbiochem)
:
127-08-2
:
:
CH₃COOK
:
MFCD00012458
:
98.14 g/mol
:
13,07687
:
304 °C
:
1.57 g/cm³ (25 °C)
:
>250 °C
:
Ambient
:
204-822-2