Order Entry
Germany
ContactUsLinkComponent
 
Potassium acetate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
Potassium acetate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Meets ACS Reagent Chemical Requirements:
    = 99.0 %
  • For Laboratory,Research,or Manufacturing Use:
    = 99.0 %
  • Assay (CH₃COOK) (by non-aqueous titration):
    ≥99.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    6.5 - 9.0
  • Chloride (Cl):
    ≤0.003 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • Sodium (Na):
    ≤0.03 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm