Order Entry
Germany
ContactUsLinkComponent
Pelargonic acid, Supelco®
Corrosive
Pelargonic acid, Supelco®
Catalog # 73982-5ML
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Pelargonic acid, Supelco®
Catalog # 73982-5ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

About this item

 :
 :158.24 g/mol
Merck Index:12,07198
Boiling Pt:253 °C (1013 hPa)
Melting Pt:12 °C
Density:0.9055 g/cm³ (20 °C)
Flash Pt:133 °C
Storage Temperature:Refrigerator
EINECS:203-931-2
UN:3265
ADR:8,III
 :M-Clarity™ quality level = MQ100
 :GHS05
CAS Number:112-05-0
Formula:C₉H₁₈O₂
MDL Number:MFCD00004433