Pelargonic acid 97%
:112-05-0
:
:C₉H₁₈O₂
:MFCD00004433
:158.24 g/mol
:12,07198
:254 °C (760 mmHg)
:12.5 °C
:0.9 g/cm³
:114 °C
:Ambient
:203-931-2
:8,III
:GHS07
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4300 to 1.4340 (20°C, 589 nm)
- GC:≥96.0 %