

Formic acid 99-100%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Boiling Pt:100…101 °C (1013 hPa)
Melting Pt:8,2…8,4 °C
Density:1.22 g/cm³ (25 °C)
Flash Pt:48 °C
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8 (3),II
REACH:01-2119491174-37
:GHS02
:GHS05
Specification Test Results
- Assay:Min. 99.0 %
- Dilution test:Passes test ACS
- Colouration:Max. 10 APHA
- Solidification point:7 - 8 °C
- Refractive index (20°C):1.370 - 1.372
- Density (20/20):1.210 - 1.230
- Acetic acid:Max. 0.2 %
- Evaporation residue:Max. 30 ppm
- Heavy metals (by ICP-MS):Max. 10 ppm
- Ignition residue (SO4):Max. 20 ppm
- Water:Max. 1.0 %
- Cl (Chloride):Max. 5 ppm
- NH4 (Ammonium):Max. 20 ppm
- SO3 (Sulphite):Max. 10 ppm
- SO4 (Sulphate):Max. 10 ppm
- Cd (Cadmium):Max. 0.1 ppm
- Cu (Copper):Max. 0.1 ppm
- Fe (Iron):Max. 4 ppm
- Pb (Lead):Max. 0.1 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test