Acetic acid glacial ≥99.0%, ULTREX® II, Ultrapure, J.T.Baker®
Catalog # 6903.0500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:= 50 ppt
- Assay (CH₃COOH):≥99.0 %
- Trace Impurities - Aluminum (Al):≤50 ppt
- Trace Impurities - Antimony (Sb):≤50 ppt
- Trace Impurities - Arsenic (As):≤50 ppt
- Trace Impurities - Barium (Ba):≤10 ppt
- Trace Impurities - Beryllium (Be):≤10 ppt
- Trace Impurities - Bismuth (Bi):≤10 ppt
- Trace Impurities - Cadmium (Cd):≤10 ppt
- Trace Impurities - Calcium (Ca):≤50 ppt
- Trace Impurities - Cerium (Ce):≤10 ppt
- Trace Impurities - Cesium (Cs):≤10 ppt
- Trace Impurities - Chromium (Cr):≤10 ppt
- Trace Impurities - Cobalt (Co):≤10 ppt
- Trace Impurities - Copper (Cu):≤10 ppt
- Trace Impurities - Dysprosium (Dy):≤1 ppt
- Trace Impurities - Erbium (Er):≤1 ppt
- Trace Impurities - Europium (Eu):≤1 ppt
- Trace Impurities - Gadolinium (Gd):≤1 ppt
- Trace Impurities - Gallium (Ga):≤10 ppt
- Trace Impurities - Germanium (Ge):≤10 ppt
- Trace Impurities - Hafnium (Hf):≤10 ppt
- Trace Impurities - Holmium (Ho):≤1 ppt
- Trace Impurities - Indium (In):≤1 ppt
- Trace Impurities - Iron (Fe):≤50 ppt
- Trace Impurities - Lanthanum (La):≤1 ppt
- Trace Impurities - Lead (Pb):≤10 ppt
- Trace Impurities - Lithium (Li):≤10 ppt
- Trace Impurities - Lutetium (Lu):≤10 ppt
- Trace Impurities - Magnesium (Mg):≤50 ppt
- Trace Impurities - Manganese (Mn):≤10 ppt
- Trace Impurities - Molybdenum (Mo):≤10 ppt
- Trace Impurities - Neodymium (Nd):≤1 ppt
- Trace Impurities - Nickel (Ni):≤50 ppt
- Trace Impurities - Platinum (Pt):≤50 ppt
- Trace Impurities - Potassium (K):≤50 ppt
- Trace Impurities - Praseodymium (Pr):≤1 ppt
- Trace Impurities - Rhenium (Re):≤10 ppt
- Trace Impurities - Rhodium (Rh):≤50 ppt
- Trace Impurities - Rubidium (Rb):≤10 ppt
- Trace Impurities - Ruthenium (Ru):≤50 ppt
- Trace Impurities - Samarium (Sm):≤1 ppt
- Trace Impurities - Scandium (Sc):≤10 ppt
- Trace Impurities - Selenium (Se), For Information Only:= 100 ppt
- Trace Impurities - Silver (Ag):≤50 ppt
- Trace Impurities - Sodium (Na):≤100 ppt
- Trace Impurities - Strontium (Sr):≤10 ppt
- Trace Impurities - Tellurium (Te):≤1 ppt
- Trace Impurities - Terbium (Tb):≤1 ppt
- Trace Impurities - Thallium (Tl):≤10 ppt
- Trace Impurities - Thorium (Th):≤1 ppt
- Trace Impurities - Thulium (Tm):≤1 ppt
- Trace Impurities - Tin (Sn):≤50 ppt
- Trace Impurities - Titanium (Ti):≤10 ppt
- Trace Impurities - Tungsten (W):≤10 ppt
- Trace Impurities - Uranium (U):≤1 ppt
- Trace Impurities - Vanadium (V):≤10 ppt
- Trace Impurities - Ytterbium (Yb):≤1 ppt
- Trace Impurities - Yttrium (Y):≤1 ppt
- Trace Impurities - Zinc (Zn):≤50 ppt
- Trace Impurities - Zirconium (Zr):≤10 ppt
Specification Test Results
About this item
:
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152