Order Entry
Germany
ContactUsLinkComponent
Acetic acid ≥96%, AnalaR NORMAPUR® analytical reagent
Acetic acid ≥96%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Acetic acid ≥96%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96.0 %
  • Substances reducing Cr6+ (as O):
    Passes test
  • Colouration:
    Max. 10 APHA
  • Evaporation residue:
    Max. 10 ppm
  • Cl (Chloride):
    Max. 1 ppm
  • SO4 (Sulphate):
    Max. 1 ppm
  • Ag (Silver):
    Max. 0.5 ppm
  • Al (Aluminium):
    Max. 0.05 ppm
  • As (Arsenic):
    Max. 0.01 ppm
  • Ba (Barium):
    Max. 0.01 ppm
  • Be (Beryllium):
    Max. 0.02 ppm
  • Bi (Bismuth):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.08 ppm
  • Cu (Copper):
    Max. 0.05 ppm
  • Fe (Iron):
    Max. 0.4 ppm
  • Ge (Germanium):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Li (Lithium):
    Max. 0.5 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Mo (Molybdenum):
    Max. 0.02 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Ni (Nickel):
    Max. 0.1 ppm
  • Pb (Lead):
    Max. 0.02 ppm
  • Sr (Strontium):
    Max. 0.01 ppm
  • Ti (Titanium):
    Max. 0.05 ppm
  • Tl (Thallium):
    Max. 0.05 ppm
  • V (Vanadium):
    Max. 0.01 ppm
  • Zn (Zinc):
    Max. 0.05 ppm
  • Zr (Zirconium):
    Max. 0.1 ppm

Specification Test Results