Order Entry
Germany
ContactUsLinkComponent
Acetic acid glacial ≥99.9%, AnalaR NORMAPUR®, for cryoscopy
Acetic acid glacial ≥99.9%, AnalaR NORMAPUR®, for cryoscopy
Catalog # 20105.292
Supplier:  VWR Chemicals
CAS Number:  
undefined
Acetic acid glacial ≥99.9%, AnalaR NORMAPUR®, for cryoscopy
Catalog # 20105.292
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99.9 %
  • Substances reducing dichromate:
    Passes test ACS
  • Colouration:
    Max. 10 APHA
  • Solidification point:
    16.3 - 16.5 °C
  • Substances coloured by H2SO4:
    Max. 150 APHA
  • Evaporation residue:
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 10 ppm
  • Water:
    Max. 0.15 %
  • Cl (Chloride):
    Max. 1 ppm
  • NO3 (Nitrate):
    Max. 1 ppm
  • SO4 (Sulphate):
    Max. 1 ppm
  • Ag (Silver):
    Max. 0.5 ppm
  • Al (Aluminium):
    Max. 0.05 ppm
  • As (Arsenic):
    Max. 0.01 ppm
  • Ba (Barium):
    Max. 0.01 ppm
  • Be (Beryllium):
    Max. 0.01 ppm
  • Bi (Bismuth):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.05 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.08 ppm
  • Cu (Copper):
    Max. 0.1 ppm
  • Fe (Iron):
    Max. 0.3 ppm
  • Ge (Germanium):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Li (Lithium):
    Max. 0.5 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Mo (Molybdenum):
    Max. 0.02 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Ni (Nickel):
    Max. 0.1 ppm
  • Pb (Lead):
    Max. 0.02 ppm
  • Sr (Strontium):
    Max. 0.01 ppm
  • Ti (Titanium):
    Max. 0.1 ppm
  • Tl (Thallium):
    Max. 0.05 ppm
  • V (Vanadium):
    Max. 0.01 ppm
  • Zn (Zinc):
    Max. 0.05 ppm
  • Zr (Zirconium):
    Max. 0.1 ppm

Specification Test Results

About this item

 :
 :60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 (3),II
REACH:01-2119475328-30
 :GHS02
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152