Order Entry
Germany
ContactUsLinkComponent
Acetic acid glacial Ph. Eur., USP
Acetic acid glacial Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates
undefined
Acetic acid glacial Ph. Eur., USP
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Appearance:
    Conforms (see CoA/CoS)
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Residual solvents:
    Passes test
  • As (Arsenic):
    Max. 1 ppm
  • Cd (Cadmium):
    Max. 0.2 ppm
  • Co (Cobalt):
    Max. 0.2 ppm
  • Hg (Mercury):
    Max. 1 ppm
  • Ni (Nickel):
    Max. 0.2 ppm
  • Pb (Lead):
    Max. 0.5 ppm
  • V (Vanadium):
    Max. 0.2 ppm
  • Conforms to Ph.Eur./USP:
    Passes test
  • Ph.Eur. specifications::
    .
  • Assay [Ph.Eur.]:
    99.0 --> 100.5 %
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Appearance test:
    Passes test Ph.Eur.
  • Freezing point [Ph.Eur.]:
    Min. 14.8 °C
  • Reducing substances:
    Passes test Ph.Eur.
  • Cl (Chloride) [Ph.Eur.]:
    Max. 25 mg/l
  • SO4 (Sulphate) [Ph.Eur.]:
    Max. 50 mg/l
  • Fe (Iron) [Ph.Eur.]:
    Max. 5 ppm
  • Residue on evaporation [Ph.Eur.]:
    Max. 0.01 %
  • USP specifications::
    .
  • Assay [USP]:
    99.5 --> 100.5 %
  • Identification:
    Passes test USP
  • Limit of nonvolatile residue [USP]:
    Max. 1.0 mg
  • Cl (Chloride):
    Passes test USP
  • SO4 (Sulphate):
    Passes test USP
  • Readily oxidisable substances:
    Passes test USP
  • Congealing temperature [USP]:
    Min. 15.6 °C

Specification Test Results