Order Entry
Germany
ContactUsLinkComponent
Acetic acid glacial 100%, EMPARTA® ACS analytical reagent, Supelco®
Acetic acid glacial 100%, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.01830.2500
Supplier:  Merck
CAS Number:  
undefined
Acetic acid glacial 100%, EMPARTA® ACS analytical reagent, Supelco®
Catalog # 1.01830.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    17 °C
  • Molar Mass:
    60.05 g/mol
  • Boiling Point:
    116 - 118 °C (1013 hPa)
  • Vapor Pressure:
    15.4 hPa (20 °C)
  • Flash Point:
    39 °C
  • Refractive Index:
    1.37 (20 °C)
  • Explosion Limit:
    4 - 19.9 %(V)
  • Density:
    1.05 g/cm3 (20 °C)
  • pH:
    2.5 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    485 °C
  • Viscosity kinematic:
    1.17 mm2/s (20 °C)
  • Evaporation number:
    24

Specification Test Results

About this item

 :
 :60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 (3),II
 :M-Clarity™ quality level = MQ200
REACH:01-2119475328-30
 :GHS02
 :GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152