Order Entry
Germany
ContactUsLinkComponent
Acetic acid glacial 99.8-100.5% (by alkalimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E260, SAFC®
Acetic acid glacial 99.8-100.5% (by alkalimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E260, SAFC®
Supplier:  Merck
Certificates
undefined
Acetic acid glacial 99.8-100.5% (by alkalimetry), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, E260, SAFC®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (alkalimetric):
    99,8 - 100,5 %
  • Identity:
    passes test
  • Appearance of the substance:
    passes test
  • Solidification temperature:
    ≥ 15,6 °C
  • Foreign organic substances:
    passes test
  • Formic acid, Formates and other oxidable impurities (as HCOOH):
    ≤ 0,10 %
  • Heavy metals (as Pb):
    ≤ 0,0005 %
  • Chloride (Cl):
    ≤ 0,0002 %
  • Sulfate (SO₄):
    ≤ 0,0005 %
  • Al (Aluminium):
    ≤ 0,00005 %
  • As (Arsenic):
    ≤ 0,00010 %
  • Cu (Copper):
    ≤ 0,0005 %
  • Fe (Iron):
    ≤ 0,0005 %
  • Hg (Mercury):
    ≤ 0,00010 %
  • Pb (Lead):
    ≤ 0,00005 %
  • Zn (Zinc):
    ≤ 0,0005 %
  • Other residual solvents (ICH Q3C):
    ...excluded by production process
  • Reducing substances (Ph Eur):
    passes test
  • Substances reducing potassium permanganate:
    passes test
  • Evaporation residue:
    ≤ 0,005 %

Specification Test Results

Related Information