Order Entry
Germany
ContactUsLinkComponent
tert-Butyl acetate for synthesis, Sigma-Aldrich®
tert-Butyl acetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
tert-Butyl acetate for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    116.15 g/mol
  • Boiling Point:
    95 - 97 °C (1013 hPa)
  • Vapor Pressure:
    41 hPa (20 °C)
  • Flash Point:
    1 °C
  • Refractive Index:
    1.387 (20 °C, 589 nm)
  • Explosion Limit:
    1.5 %(V)
  • Density:
    0.87 g/cm3 (20 °C)

Specification Test Results

Related Information