sec-Butyl acetate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105-46-4
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009328
:116.16 g/mol
Merck Index:14,1536
Boiling Pt:111…112 °C
Melting Pt:–99 °C
Density:0.872
Flash Pt:16 °C (60 °F)
Storage Temperature:Ambient
EINECS:203-300-1
UN:1123
ADR:3,III
:GHS02