Order Entry
Germany
ContactUsLinkComponent
1,3-Dihydroxyacetone for synthesis, Sigma-Aldrich®
1,3-Dihydroxyacetone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,3-Dihydroxyacetone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    250 g/l (20 °C)
  • Melting Point:
    83 °C Monomer
  • Molar Mass:
    90.07 g/mol
  • Boiling Point:
    188 °C (decomposition),OECD Test Guideline 103
  • Vapor Pressure:
     - 0.01 Pa (20 °C)
  • Density:
    1.52 g/cm3 (20 °C) OECD Test Guideline 109
  • pH:
    4 - 6 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number