Order Entry
Germany
ContactUsLinkComponent
Acetone ≥99.0% Ph. Eur.
Acetone ≥99.0% Ph. Eur.
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Acetone ≥99.0% Ph. Eur.
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Appearance:
    Clear colourless liquid
  • Identification B:
    Passes test
  • Identification C:
    Passes test
  • Appearance of solution:
    Passes test
  • Acidity or alkalinity:
    Passes test
  • Relative density:
    0.79 - 0.793
  • Reducing substances:
    Passes test
  • Methanol (V/V):
    Max. 0.05 %
  • Propan-2-ol (V/V):
    Max. 0.05 %
  • Benzene (V/V):
    Max. 2 ppm
  • Any other impurity (V/V):
    Max. 0.05 %
  • Matter insoluble in water:
    Passes test
  • Residue on evaporation:
    Max. 50 ppm
  • Water:
    Max. 3 g/l
  • Residual solvents:
    Passes test
  • Mesityl oxide (*):
    Max. 10 ppm
  • Statement of original manufacturer:
    (*)

Specification Test Results