Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Acetone ≥99%, TECHNICAL

Flammable
Irritant
   Sustainable Options Available
Supplier:   VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
Acetone ≥99%, TECHNICAL

6 Options Available Below

Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test

Specifications

Specifications

  • Catalog No:
  • 20063.296
  • 20063.365
  • 20063.412
  • 20063.467
  • 20063.558
  • 20063.930
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 21,80 €
    21,80 €
    1 * 1 L
       21,80 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20063.296
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic container
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 72,50 €
    72,50 €
    1 * 5 L
       72,50 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20063.365
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic container
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 152,00 €
    152,00 €
    1 * 10 L
       152,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20063.412
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic drum
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 235,00 €
    235,00 €
    1 * 25 L
       235,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20063.467
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Metal drum
    200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L - 1 410,00 €
    1 410,00 €
    1 * 200 L
       1 410,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20063.558
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 950 L
    Log in for availability
    Log in for availability
    1 * 950 L
    1 * 950 L

    Specifications:
    More
    Specifications
    Cat. No.
    20063.930
    Environmentally Preferable
    Environmental certification