Order Entry
Germany
Orders LinkContactUsLinkComponent
Acetone analytical reagent

Acetone analytical reagent

Flammable
Irritant
   Sustainable Options Available
Supplier:   Bernd Kraft
Acetone analytical reagent
Supplier:  Bernd Kraft
   Sustainable Options Available
Certificates
Acetone analytical reagent

8 Options Available Below

Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • KRAF07021.2000
  • KRAF07021.4000
  • KRAF07021.4700
  • KRAF07021.3000
  • KRAF07021.3700
  • KRAF07021.5300
  • KRAF07021.8900
  • KRAF07021.8500
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.2000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 64,80 €
    64,80 €
    1 * 2.500 mL
       64,80 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.4000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 66,70 €
    66,70 €
    1 * 2.500 mL
       66,70 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.4700
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 36,30 €
    36,30 €
    1 * 1 L
       36,30 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.3000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 38,20 €
    38,20 €
    1 * 1 L
       38,20 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.3700
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 117,00 €
    117,00 €
    1 * 5 L
       117,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.5300
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 381,00 €
    381,00 €
    1 * 25 L
       381,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.8900
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • 25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L - 357,00 €
    357,00 €
    1 * 25 L
       357,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07021.8500
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.