Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Supplier:   Brand
Watch glasses
Supplier:  Brand
Certificates
Watch glasses
6 Options Available Below
Product Details & Documents
PTFE, white.
  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth surfaces to prevent contamination
Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.
Product Family Options
Product Information
  • Ø
    Availability
    Price
  • BRND150904
    Watch glass, PTFE
    Watch glass, PTFE
    40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,85 €
    9,85 €
    1 * 1 ST
       9,85 €
  • BRND150907
    Watch glass, PTFE
    Watch glass, PTFE
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,10 €
    16,10 €
    1 * 1 ST
       16,10 €
  • 216-1155
    Watch glass, PTFE
    Watch glass, PTFE
    75 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,85 €
    18,85 €
    1 * 1 ST
       18,85 €
  • BRND150912
    Watch glass, PTFE
    Watch glass, PTFE
    80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,60 €
    21,60 €
    1 * 1 ST
       21,60 €
  • BRND150915
    Watch glass, PTFE
    Watch glass, PTFE
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • BRND150920
    Watch glass, PTFE
    Watch glass, PTFE
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,10 €
    38,10 €
    1 * 1 ST
       38,10 €