Order Entry
Germany
ContactUsLinkComponent
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.5 %
  • Aldehyde (as HCHO):
    max. 0.002 %
  • Color (APHA):
    max. 10
  • Diisopropylether((CH3)2CHOCH(CH3)2):
    max. 0.05 %
  • Isopropyl Alcohol:
    max. 0.05 %
  • Methanol (CH3OH):
    max. 0.05 %
  • Residue after Evaporation:
    max. 5 ppm
  • Solubility in Water:
    passes test
  • Substances Reducing KMnO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0.0003
  • Titrable Base (meq/g):
    max. 0.0006
  • Water (H2O):
    max. 0.5 %
  • Aluminium (Al):
    max. 0.5
  • Boron (B):
    max. 0.02
  • Barium (Ba):
    max. 0.1
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.05
  • Cobalt (Co):
    max. 0.02
  • Chromium (Cr):
    max. 0.02
  • Copper (Cu):
    max. 0.02
  • Iron (Fe):
    max. 0.1
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.02
  • Nickel (Ni):
    max. 0.02
  • Lead (Pb):
    max. 0.1
  • Tin (Sn):
    max. 0.1
  • Zinc (Zn):
    max. 0.1
  • at 330 nm:
    max. 1.00
  • at 340 nm:
    max. 0.10
  • at 350 nm:
    max. 0.02
  • at 400 nm:
    max. 0.01

Specification Test Results

Related Information