Acetone ≥99.5% for spectrophotometry
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% for spectrophotometry
Supplier: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
Purity: 99.5+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765
Synonyme: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS-Nummer:67-64-1
- Formel::CH₃COCH₃
- MDL-Nummer:MFCD00008765
- Molekulargewicht:58.08 g/mol
- Merck-Index:14,66
- Siedepunkt:56 °C
- Schmelzpunkt:–94 °C
- Dichte:0.791
- Flammpunkt:–17 °C (1 °F)
- Lagertemperatur:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
2 mögliche Produktvariationen vorhanden
Product Details & Documents
Purity: 99.5+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765
Synonyme: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS-Nummer:67-64-1
- Formel::CH₃COCH₃
- MDL-Nummer:MFCD00008765
- Molekulargewicht:58.08 g/mol
- Merck-Index:14,66
- Siedepunkt:56 °C
- Schmelzpunkt:–94 °C
- Dichte:0.791
- Flammpunkt:–17 °C (1 °F)
- Lagertemperatur:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Spezifikationen für diese Produktfamilie
Spezifikationen:
Spezifikationen:
- Catalog No:
- 32451.K2
- 32451.M1
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
Spezifikationen:
Product Family Options
Produktinformationen
- Pack typePk InfoPkAvailabilityPrice
- Spezifikationen:
Safer Human and Environmental Health More
Specifications
Cat. No.32451.K2Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health - Spezifikationen:
Safer Human and Environmental Health More
Specifications
Cat. No.32451.M1Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health



